Antimony ≥99.9999% (metals basis), shot, Puratronic® 1.5-5 mm (0.06-0.2 in)
Supplier: Thermo Fisher Scientific
Warning
00428.06 00428.14 00428.22
00428.06EA 55.6 EUR
00428.06 00428.14 00428.22
Antimony ≥99.9999% (metals basis), shot, Puratronic® 1.5-5 mm (0.06-0.2 in)
Antimony
Purity: 99.9999% CAS No.: 7440-36-0 Odour: Odourless MDL No.: MFCD00134030
Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637
Melting Pt: 630
Density: 6.684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733