Antimony ≥99% (metals basis), shot ≤ 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Warning
45095.22 45095.A1 45095.36
45095.22EA 50.7 EUR
45095.22 45095.A1 45095.36
Antimony ≥99% (metals basis), shot ≤ 6.35 mm (0.25 in)
Antimony
Purity: 99% CAS No.: 7440-36-0 Odour: Odourless MDL No.: MFCD00134030
Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637
Melting Pt: 630
Density: 6.684 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733