Arsenic ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Supplier: Thermo Scientific
Danger
Synonyms: Arsenic solution
ampouled under argon
13375.09 13375.14 13375.22 13375.A1
13375.09EA 116 EUR
13375.09 13375.14 13375.22 13375.A1
Arsenic ≥99.9999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Arsenic
Purity: 99.9999% CAS No.: 7440-38-2 Physical Form: Ampouled under argon Odour: Odourless UN No.: 1558 MDL No.: MFCD00085309
Formula: As
MW: 74.92 g/mol
Melting Pt: 817 °C
Density: 5.727 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085309
CAS Number: 7440-38-2
EINECS: 213-148-6
UN: 1558
ADR: 6.1,II
Merck Index: 12,00832