Bismuth ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
00915.30 00915.A1
00915.30EA 219 EUR
00915.30 00915.A1
Bismuth ≥99.9% (metals basis), rods, Ø 12.7 mm (0.5 in)
Bismuth
Purity: 99.9% CAS No.: 7440-69-9 Physical Form: ≈12.5g/cm Odour: Odourless MDL No.: MFCD00134033
Formula: Bi
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297