Bismuth ≥99.998% (metals basis), needles 1-5 cm, Length: 1-5 cm (0.4-2 in)
Supplier: Thermo Fisher Scientific
10109.22 10109.36 10109.A3
10109.22EA 84 EUR
10109.22 10109.36 10109.A3
Bismuth ≥99.998% (metals basis), needles 1-5 cm, Length: 1-5 cm (0.4-2 in)
Bismuth
Purity: 99.998% CAS No.: 7440-69-9 Physical Form: Approximately 1.5mm diameter Odour: Odourless MDL No.: MFCD00134033
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297