Bismuth ≥99.99% (metals basis), pieces ≤ 10 cm (3.9 in)
Supplier: Thermo Fisher Scientific
12208.30 12208.A1 12208.A7
12208.30EA 112 EUR
12208.30 12208.A1 12208.A7
Bismuth ≥99.99% (metals basis), pieces ≤ 10 cm (3.9 in)
Bismuth
Purity: 99.99% CAS No.: 7440-69-9 Odour: Odourless MDL No.: MFCD00134033
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297