Bismuth ≥99.999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Supplier: Thermo Fisher Scientific
14442.22 14442.36 14442.A3
14442.22EA 80.7 EUR
14442.22 14442.36 14442.A3
Bismuth ≥99.999% (metals basis), polycrystalline lumps, Puratronic® 2-8 mm (0.08-0.3 in)
Bismuth
Purity: 99.999% CAS No.: 7440-69-9 Odour: Odourless MDL No.: MFCD00134033
Formula: Bi
MW: 208.98 g/mol
Boiling Pt: 1490
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297