Bismuth ≥99.5% (metals basis), powder -100 mesh
Supplier: Thermo Fisher Scientific
Warning
10110.22 10110.36 10110.A3
10110.22EA 66.9 EUR
10110.22 10110.36 10110.A3
Bismuth ≥99.5% (metals basis), powder -100 mesh
Bismuth
Purity: 99.5% CAS No.: 7440-69-9 Physical Form: Apparent density 3.8-4.6g/cm3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00134033
Formula: Bi
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
UN: 3089
ADR: 4.1,III