Sulphur ≥99.5%, powder, precipitated
Supplier: Thermo Fisher Scientific
Danger
10785.36 10785.A3
10785.36EA 61.2 EUR
10785.36 10785.A3
Sulphur ≥99.5%, powder, precipitated
Sulphur
Purity: 99.5% CAS No.: 7704-34-9 Odour: Weak UN No.: 1350 MDL No.: MFCD00085316
Formula: S
MW: 32.07 g/mol
Boiling Pt: 445
Melting Pt: 113…119 °C
Density: 2.36 g/cm³ (20 °C)
Flash Pt: 168
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1 III
Merck Index: 13,09059