Sulphur, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
213292-10G
213292-250G
213292-50G
SIAL213292-10GEA
62.6
EUR
SIAL213292-10G
SIAL213292-250G
SIAL213292-50G
Sulphur, Sigma-Aldrich®
Sulphur
Formula:
S MW: 32.07 g/mol Boiling Pt: 445 Melting Pt: 113…119 °C Density: 2.36 g/cm³ (20 °C) Flash Pt: 168 Storage Temperature: Ambient |
MDL Number:
MFCD00085316 CAS Number: 7704-34-9 EINECS: 231-722-6 UN: 1350 ADR: 4.1 III Merck Index: 13,09059 |