Sulphur, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL414980-50GEA
126
EUR
SIAL414980-50G
SIAL414980-250G
Sulphur, Sigma-Aldrich®
Sulphur
Formula:
S MW: 32,07 g/mol Boiling Pt: 445 °C (1013 hPa) Melting Pt: 113…119 °C Density: 2,36 g/cm³ (20 °C) Flash Pt: 168 °C |
MDL Number:
MFCD00085316 UN: 1350 ADR: 4.1,III Merck Index: 13,09059 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...