Tellurium, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
452378-25G
452378-5G
SIAL452378-25GEA
446
EUR
SIAL452378-25G
SIAL452378-5G
Tellurium, Sigma-Aldrich®
Tellurium
Formula:
Te Boiling Pt: 990 °C (1013 hPa) Melting Pt: 450 °C Density: 8.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3284 ADR: 6.1,III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...