Erbium, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL263044-5GEA 501 EUR
SIAL263044-5G
Erbium, powder, Sigma-Aldrich®
Erbium
Formula: Er
MW: 167,26 g/mol
Boiling Pt: 2863 °C (1013 hPa)
Melting Pt: 1529 °C
Density: 9,066 g/cm³ (20 °C)
UN: 3178
ADR: 4.1,II
Merck Index: 12,03679

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR