Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Supplier: Thermo Scientific
Packaged under argon.
00595.FF 00595.FI
00595.FFEA 200 EUR
00595.FF 00595.FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Gadolinium
Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈3.1g/25x25mm, Packaged under argon MDL No.: MFCD00011022
Formula: Gd
MW: 157.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2