Neodymium, powder, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

SIAL460877-10GEA 220 EUR
SIAL460877-10G
Neodymium, powder, Sigma-Aldrich®
Neodymium
Formula: Nd
MW: 144,24 g/mol
Boiling Pt: 2474 °C (1013 hPa)
Melting Pt: 1014,85 °C
Density: 6,46 g/cm³ (1240 °C)
UN: 3178
ADR: 4.1,III
Merck Index: 12,06538

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR