Gold 99,99 Metals basis, foil, Premion®, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
00132.FF 00132.FI
00132.FFEA 535 EUR
00132.FF 00132.FI
Gold 99,99 Metals basis, foil, Premion®, Thickness: 0.127 mm (0.005 in)
Gold
Purity: 99.99% CAS No.: 7440-57-5 Physical Form: ≈1.5g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526