Gold ≥99.985% (metals basis), foil, Premion®, Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Fisher Scientific
00133.FI 00133.FF
00133.FIEA 621 EUR
00133.FI 00133.FF
Gold ≥99.985% (metals basis), foil, Premion®, Thickness: 0.025 mm (0.001 in)
Gold
Purity: 99.985% CAS No.: 7440-57-5 Physical Form: ≈0.3g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526