Gold ≥99.995% (metals basis), powder, Premion® -20/+100 mesh
Supplier: Thermo Fisher Scientific
00136.03 00136.06
00136.03EA 343 EUR
00136.03 00136.06
Gold ≥99.995% (metals basis), powder, Premion® -20/+100 mesh
Gold
Purity: 99.995% CAS No.: 7440-57-5 Physical Form: Agglomerated MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526