Gold 99,9975 Metals basis, foil, Premion®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
11390.FF 11390.FI 11390.GH
11390.FFEA 964 EUR
11390.FF 11390.FI 11390.GH
Gold 99,9975 Metals basis, foil, Premion®, Thickness: 0.25 mm (0.01 in)
Gold
Purity: 99.9975+% CAS No.: 7440-57-5 Physical Form: ≈3g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526