Gold ≥99.9% (metals basis), thin foil, Thickness: 0.01 mm (0.0004 in)
Supplier: Thermo Fisher Scientific
11518.FF 11518.FI 11518.GH
11518.FFEA 256 EUR
11518.FF 11518.FI 11518.GH
Gold ≥99.9% (metals basis), thin foil, Thickness: 0.01 mm (0.0004 in)
Gold
Purity: 99.9% CAS No.: 7440-57-5 Physical Form: ≈0.12g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526