Gold ≥99.9985% (metals basis), rods, Premion®, Ø 3 mm (0.1 in)
Supplier: Thermo Fisher Scientific
11892.06 11892.93
11892.06EA 1530 EUR
11892.06 11892.93
Gold ≥99.9985% (metals basis), rods, Premion®, Ø 3 mm (0.1 in)
Gold
Purity: 99.9985% CAS No.: 7440-57-5 Physical Form: ≈3.7cm/5g Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526