Gold ≥99.95% (metals basis), flake/sphere APS 1-15 µm
Supplier: Thermo Fisher Scientific
12453.03 12453.06
12453.03EA 322 EUR
12453.03 12453.06
Gold ≥99.95% (metals basis), flake/sphere APS 1-15 µm
Gold
Purity: 99.95% CAS No.: 7440-57-5 Physical Form: S.A. 0.24-0.34m2/g MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526