Gold 99,99 Metals basis, slug, Premion®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
13394.KG 13394.KJ 13394.KM
13394.KGEA 312 EUR
13394.KG 13394.KJ 13394.KM
Gold 99,99 Metals basis, slug, Premion®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Gold
Purity: 99.99% CAS No.: 7440-57-5 Physical Form: ≈0.97g/slug Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526