Gold 99,95 Metals basis, wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
14724.BU 14724.BY 14724.CC
14724.BUEA 120 EUR
14724.BU 14724.BY 14724.CC
Gold 99,95 Metals basis, wire, Ø 0.25 mm (0.01 in)
Gold
Purity: 99.95% CAS No.: 7440-57-5 Physical Form: ≈0.95g/m Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526