Gold 14kt (Au ≥ 58,3%), yellow, wire, Ø 1.63 mm (0.064 in)
Supplier: Thermo Fisher Scientific
40329.BS 40329.BU
40329.BSEA 531 EUR
40329.BS 40329.BU
Gold 14kt (Au ≥ 58,3%), yellow, wire, Ø 1.63 mm (0.064 in)
Gold
Molecular Formula: % Concentration CuZnAg Physical Form: ≈28g/m Appearance: Yellow MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526