Gold 99,9 Metals basis, foil_thin, Thickness: 0.005 mm (0.0002 in)
Supplier: Thermo Fisher Scientific
40739.FF 40739.FI
40739.FFEA 565 EUR
40739.FF 40739.FI
Gold 99,9 Metals basis, foil_thin, Thickness: 0.005 mm (0.0002 in)
Gold
Purity: 99.9% CAS No.: 7440-57-5 Physical Form: ≈0.06g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526