Gold 99,96 Metals basis, flake/sphere APS 2,2-3,5 µm
Supplier: Thermo Fisher Scientific
42153.03 42153.06
42153.03EA 322 EUR
42153.03 42153.06
Gold 99,96 Metals basis, flake/sphere APS 2,2-3,5 µm
Gold
Purity: 99.96% CAS No.: 7440-57-5 Physical Form: Thick flake MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526