Gold ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
43071.GH
43071.FF
43071.FI
43071.GHEA
2750
EUR
43071.GH
43071.FF
43071.FI
Gold ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Gold
Purity: 99.95%
CAS No.: 7440-57-5
Physical Form: ≈0.6g/25x25mm
Odour: Odourless
MDL No.: MFCD00003436
Formula:
Au Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |