Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.35 mm (0.0138 in)
Supplier: Thermo Fisher Scientific
45084.CC 45084.BY 45084.BU
45084.CCEA 2630 EUR
45084.CC 45084.BY 45084.BU
Gold ≥99.999% (metals basis), wire, Premion®, Ø 0.35 mm (0.0138 in)
Gold
Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈1.82g/m Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526