Gold, Sigma-Aldrich®
Supplier: Merck
MQ100
265799-7.5G
265799-750MG
SIAL265799-7.5GEA
1680
EUR
SIAL265799-7.5G
SIAL265799-750MG
Gold, Sigma-Aldrich®
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...