Gold, Sigma-Aldrich®
Supplier: Merck
MQ100
326496-1.5G
326496-6G
SIAL326496-1.5GEA
658
EUR
SIAL326496-1.5G
SIAL326496-6G
Gold, Sigma-Aldrich®
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 Melting Pt: 1063 Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |