Gold, Sigma-Aldrich®

Supplier: Merck


MQ100

326534-2.8G 326534-280MG
SIAL326534-2.8GEA 680 EUR
SIAL326534-2.8G SIAL326534-280MG
Gold, Sigma-Aldrich®
Gold
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR