Gold, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
349275-2.4G
349275-600MG
SIAL349275-2.4GEA
923
EUR
SIAL349275-2.4G
SIAL349275-600MG
Gold, Sigma-Aldrich®
Gold
Formula:
Au Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...