Copper 99,9995 Metals basis, discs, Ø 9.0 +/- 0.127 mm (0.354 +/- 0.005 in), Thickness: 0.8+/-0.051 mm (0.03+/-0.002 in)
Supplier: Thermo Fisher Scientific
10951.09 10951.18
10951.09EA 173 EUR
10951.09 10951.18
Copper 99,9995 Metals basis, discs, Ø 9.0 +/- 0.127 mm (0.354 +/- 0.005 in), Thickness: 0.8+/-0.051 mm (0.03+/-0.002 in)
Copper
Purity: 99.9995% CAS No.: 7440-50-8 Physical Form: ≈0.5g/disc Odour: Odourless MDL No.: MFCD00010965
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545