Silver ≥99.9985% (metals basis), rods, Premion®, Ø 12.5 mm (0.492 in)
Supplier: Thermo Fisher Scientific
11356.BP 11356.BS
11356.BPEA 180 EUR
11356.BP 11356.BS
Silver ≥99.9985% (metals basis), rods, Premion®, Ø 12.5 mm (0.492 in)
Silver
Purity: 99.9985% CAS No.: 7440-22-4 Physical Form: ≈12.87g/cm Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647