Silver 99,999 Metals basis, shots, Premion® 1-7 mm (0.04-0.1 in)
Supplier: Thermo Fisher Scientific
11357.09 11357.18 11357.30
11357.09EA 143 EUR
11357.09 11357.18 11357.30
Silver 99,999 Metals basis, shots, Premion® 1-7 mm (0.04-0.1 in)
Silver
Purity: 99.999% CAS No.: 7440-22-4 Odour: Characteristic MDL No.: MFCD00003397
For vapor deposition
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647