Silver hard ≥99.9% (metals basis), wire 1/2 hard, hard, Ø 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
11433.G2 11433.G6 11433.H2
11433.G2EA 76.8 EUR
11433.G2 11433.G6 11433.H2
Silver hard ≥99.9% (metals basis), wire 1/2 hard, hard, Ø 0.5 mm (0.02 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈2.06g/m, Temper: 1/2 hard Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647