Silver ≥99.9% (metals basis), shot 1-5 mm (0.04-0.2 in)
Supplier: Thermo Fisher Scientific
11436.09 11436.18 11436.30
11436.09EA 88.6 EUR
11436.09 11436.18 11436.30
Silver ≥99.9% (metals basis), shot 1-5 mm (0.04-0.2 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647