Silver ≥99.99% (metals basis), wire, Premion®, Ø 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
11467.G5 11467.H2
11467.G5EA 470 EUR
11467.G5 11467.H2
Silver ≥99.99% (metals basis), wire, Premion®, Ø 0.05 mm (0.002 in)
Silver
Purity: 99.99% CAS No.: 7440-22-4 Physical Form: ≈0.02g/m Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647