Silver 99,999 Metals basis, needles
Supplier: Thermo Fisher Scientific
39697.09 39697.18
39697.09EA 97.4 EUR
39697.09 39697.18
Silver 99,999 Metals basis, needles
Silver
Purity: 99.999% CAS No.: 7440-22-4 Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647