Silver, gauze_wire 50 mesh, Ø 0.0764 mm (0.003 in)
Supplier: Thermo Fisher Scientific
40935.FL 40935.GT 40935.HB
40935.FLEA 295 EUR
40935.FL 40935.GT 40935.HB
Silver, gauze_wire 50 mesh, Ø 0.0764 mm (0.003 in)
Silver
CAS No.: 7440-22-4 Physical Form: Wire Cloth Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647