Silver ≥99.9% (metals basis), wire, hard, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
41456.G1 41456.G5 41456.G9
41456.G1EA 96.5 EUR
41456.G1 41456.G5 41456.G9
Silver ≥99.9% (metals basis), wire, hard, Ø 1.0 mm (0.04 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈8.24g/m, Temper: hard Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647