Silver ≥99.999% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Fisher Scientific
42984.06 42984.14 42984.22
42984.06EA 146 EUR
42984.06 42984.14 42984.22
Silver ≥99.999% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Silver
Purity: 99.999% CAS No.: 7440-22-4 Physical Form: ≈0.26g/slug Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647