Silver ≥99.99% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
44461.G5 44461.H2 44461.J4
44461.G5EA 75.6 EUR
44461.G5 44461.H2 44461.J4
Silver ≥99.99% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)
Silver
Purity: 99.99% CAS No.: 7440-22-4 Physical Form: ≈0.02g/m Odour: Characteristic MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647