Silver 99 Metals basis, powder -120+325 mesh, atomised
Supplier: Thermo Fisher Scientific
45097.22 45097.14
45097.22EA 543 EUR
45097.22 45097.14
Silver 99 Metals basis, powder -120+325 mesh, atomised
Silver
Purity: 99% CAS No.: 7440-22-4 Physical Form: atomized powder, up to 10% variance on size Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647