Silver, Sigma-Aldrich®

Supplier: Merck


MQ100

267430-33G 267430-6.6G
SIAL267430-33GEA 422 EUR
SIAL267430-33G SIAL267430-6.6G
Silver, Sigma-Aldrich®
Silver
Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR