Silver, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

327050-10G 327050-50G
SIAL327050-10GEA 180 EUR
SIAL327050-10G SIAL327050-50G
Silver, Sigma-Aldrich®
Silver
Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR