Cadmium ≥99.999% (metals basis), wire, Puratronic®, Ø 0.64 mm (0.025 in)
Supplier: Thermo Fisher Scientific
Danger
00060.G4 00060.G7
00060.G4EA 162 EUR
00060.G4 00060.G7
Cadmium ≥99.999% (metals basis), wire, Puratronic®, Ø 0.64 mm (0.025 in)
Cadmium
Purity: 99.999% CAS No.: 7440-43-9 Physical Form: ≈2.8g/m MDL No.: MFCD00010914
Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649