Cadmium ≥99.999% (metals basis), rods, Puratronic® 23 cm (9.0 in), Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
Danger
10120.KG 10120.KJ 10120.KM
10120.KGEA 242 EUR
10120.KG 10120.KJ 10120.KM
Cadmium ≥99.999% (metals basis), rods, Puratronic® 23 cm (9.0 in), Ø 12.7 mm (0.5 in)
Cadmium
Purity: 99.999+% CAS No.: 7440-43-9 Physical Form: ≈250g/rod MDL No.: MFCD00010914
Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649