Cadmium 99,999 Metals basis, shots_tear drop 3 mm (0.1 in)
Supplier: Thermo Fisher Scientific
Danger
35483.18 35483.30
35483.18EA 149 EUR
35483.18 35483.30
Cadmium 99,999 Metals basis, shots_tear drop 3 mm (0.1 in)
Cadmium
Purity: 99.999% CAS No.: 7440-43-9 MDL No.: MFCD00010914
Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765
Melting Pt: 321
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649